2,2-二甲基-1,3-丙二醇 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
NPG Glycol, 新戊二醇
D0791-25G
D0791-500G
TCIAD0791-25GEA
0
CNY
TCIAD0791-25G
TCIAD0791-500G
2,2-二甲基-1,3-丙二醇 ≥98.0% (by GC)
2,2-二甲基-1,3-丙二醇
分子式:
C₅H₁₂O₂ 沸点: 210°C 溶点: 128…130 °C 密度: 1,07 g/cm³ (20 °C) 存储条件: Ambient |
MDL编号:
MFCD00004685 CAS号: 126-30-7 EINECS: 204-781-0 Merck Index: 14,06458 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 127.0 to 130.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...