己酸酐 ≥96.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
正己酸酐
C0021-25ML
C0021-500ML
TCIAC0021-25MLEA
0
CNY
TCIAC0021-25ML
TCIAC0021-500ML
己酸酐 ≥96.0% (by GC)
己酸酐
分子式:
C₁₂H₂₂O₃ 沸点: 244°C 溶点: –40 °C 密度: 0,93 g/cm³ (20 °C) 闪点: 128°C 存储条件: Ambient |
MDL编号:
MFCD00009509 CAS号: 2051-49-2 EINECS: 218-121-4 ADR: 8,II |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Purity (Morpholine Method) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...