羟乙基乙二胺 ≥99.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: N-(2-氨基乙基)乙醇胺, 氨基乙基乙醇胺

A0299-500ML A0299-25ML
TCIAA0299-500MLEA 0 CNY
TCIAA0299-500ML TCIAA0299-25ML
羟乙基乙二胺 ≥99.0% (by GC, titration analysis)
羟乙基乙二胺
分子式: C₄H₁₂N₂O
分子量: 104.15 g/mol
沸点: 238
溶点: -28
闪点: 125°C
存储条件: Ambient
MDL编号: MFCD00008170
CAS号: 111-41-1
EINECS: 203-867-5
UN号: 2735
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1120f1c6997fef1fd69a4bfc045c3481d9a040f1&vid=1120f1c6997fef1fd69a4bfc045c3481d9a040f1&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR