1,3-二氨基-2-丙醇 ≥97.0% (by titrimetric analysis)

供应商: TCI
Danger

Synonyms: 1,3-二氨基-2-羟基丙烷

H0497-25G H0497-5G
TCIAH0497-25GEA 0 CNY
TCIAH0497-25G TCIAH0497-5G
1,3-二氨基-2-丙醇 ≥97.0% (by titrimetric analysis)
1,3-二氨基-2-丙醇
分子式: C₃H₁₀N₂O
沸点: 114°C
溶点: 42°C
存储条件: Refrigerator
MDL编号: MFCD00008142
CAS号: 616-29-5
EINECS: 210-474-2
UN号: 3259
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=652a17f787ee7676b747d15c038de2ce3acbaa7e&vid=652a17f787ee7676b747d15c038de2ce3acbaa7e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR