三氟甲烷磺酰氯 ≥98.0% (by GC)

供应商: TCI
Danger

T1027-25G T1027-5G
TCIAT1027-25GEA 0 CNY
TCIAT1027-25G TCIAT1027-5G
三氟甲烷磺酰氯 ≥98.0% (by GC)
三氟甲烷磺酰氯
分子式: CClF₃O₂S
沸点: 31°C
密度: 1,583 g/cm³ (20 °C)
存储条件: Refrigerator
MDL编号: MFCD00007451
CAS号: 421-83-0
EINECS: 207-009-0
UN号: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=61a4804d5cdbd3fb44e1198f62f800c9818f1c42&vid=61a4804d5cdbd3fb44e1198f62f800c9818f1c42&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR