三氟乙酸酐 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: TFAA

T0433-20ML T0433-400ML
TCIAT0433-20MLEA 0 CNY
TCIAT0433-20ML TCIAT0433-400ML
三氟乙酸酐 ≥98.0% (by GC)
三氟乙酸酐
分子式: C₄F₆O₃
分子量: 210.03 g/mol
沸点: 40°C
溶点: -65
MDL编号: MFCD00000416
CAS号: 407-25-0
EINECS: 206-982-9
UN号: 3265
ADR: 8 I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=453f87c60a47788082c750b91c7b8b481732e07c&vid=453f87c60a47788082c750b91c7b8b481732e07c&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR