三氟乙酸甲酯 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: Trifluoroacetic methyl ester

T0680-25G T0680-500G
TCIAT0680-25GEA 0 CNY
TCIAT0680-25G TCIAT0680-500G
三氟乙酸甲酯 ≥98.0% (by GC)
三氟乙酸甲酯
分子式: C₃H₃F₃O₂
分子量: 128.05 g/mol
沸点: 43°C
溶点: -78
闪点: –20°C
MDL编号: MFCD00000417
CAS号: 431-47-0
EINECS: 207-074-5
UN号: 2924
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eacf298fad3aa001436f25170a0201eae9145f75&vid=eacf298fad3aa001436f25170a0201eae9145f75&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR