二氟乙酸乙酯 ≥98.0% (by GC)
供应商: TCI
|
Danger
|
Synonyms:
Difluoroacetic ethyl ester
D2498-25G
D2498-5G
TCIAD2498-25GEA
0
CNY
TCIAD2498-25G
TCIAD2498-5G
二氟乙酸乙酯 ≥98.0% (by GC)
二氟乙酸乙酯
分子式:
C₄H₆F₂O₂ 沸点: 98°C 密度: 1,1893 g/cm³ (9.8 °C) 闪点: 24°C |
MDL编号:
MFCD00013578 CAS号: 454-31-9 EINECS: 207-223-4 UN号: 2924 ADR: 3,III |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...