癸二腈 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: 1,8-亚辛基二氰化物, Decanedinitrile

S0029-25ML S0029-500ML
TCIAS0029-25MLEA 0 CNY
TCIAS0029-25ML TCIAS0029-500ML
癸二腈 ≥98.0% (by GC)
癸二腈
分子式: C₁₀H₁₆N₂
分子量: 164.25 g/mol
沸点: 200°C
MDL编号: MFCD00019900
CAS号: 1871-96-1
EINECS: 217-494-0
UN号: 3276
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f68d3c31338d8c6d33a82f3490b9fb25519dd197&vid=f68d3c31338d8c6d33a82f3490b9fb25519dd197&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR