二氟氯乙酸乙酯 ≥97.0% (by GC)
供应商: TCI
|
Danger
|
Synonyms:
Ethyl 2-chloro-2,2-difluoroacetate, Chlorodifluoroacetic ethyl ester
C1458-25G
C1458-500G
TCIAC1458-25GEA
0
CNY
TCIAC1458-25G
TCIAC1458-500G
二氟氯乙酸乙酯 ≥97.0% (by GC)
二氟氯乙酸乙酯
分子式:
C₄H₅ClF₂O₂ 沸点: 96°C 密度: 1,253 g/cm³ (20 °C) |
MDL编号:
MFCD00013662 CAS号: 383-62-0 EINECS: 206-850-0 UN号: 2924 ADR: 3,II |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...