二氟溴乙酸乙酯 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: Ethyl 2-bromo-2,2-difluoroacetate, Bromodifluoroacetic ethyl ester

B1463-25G B1463-250G B1463-5G
TCIAB1463-25GEA 0 CNY
TCIAB1463-25G TCIAB1463-250G TCIAB1463-5G
二氟溴乙酸乙酯 ≥98.0% (by GC)
二氟溴乙酸乙酯
分子式: BrF₂CCOOCH₂CH₃
沸点: 112 °C (1013 hPa)
密度: 1,5926 g/cm³ (20 °C)
闪点: 21°C
MDL编号: MFCD00042069
CAS号: 667-27-6
EINECS: 211-567-0
UN号: 2924
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1abf3b23cf648df1cdfa1455740619ff5814b16d&vid=1abf3b23cf648df1cdfa1455740619ff5814b16d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR