1-苯基-1H-四唑-5-硫醇 ≥98.0% (by HPLC, titration analysis)
供应商: TCI
|
Danger
|
Synonyms:
5-巯基-1-苯基四氮唑, 1-苯基四氮唑-5-硫醇, 1-苯基-5-巯基四氮唑
P0640-25G
P0640-500G
TCIAP0640-25GEA
0
CNY
TCIAP0640-25G
TCIAP0640-500G
1-苯基-1H-四唑-5-硫醇 ≥98.0% (by HPLC, titration analysis)
1-苯基-1H-四唑-5-硫醇
分子式:
C₇H₆N₄S 分子量: 178.22 g/mol 存储条件: Ambient |
MDL编号:
MFCD00003129 CAS号: 86-93-1 EINECS: 201-710-5 UN号: 1325 ADR: 4.1 II |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...