(S)-(+)-2-氨基-3-甲基-1-丁醇 ≥97.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: L-缬氨醇

V0058-25G V0058-5G
TCIAV0058-25GEA 0 CNY
TCIAV0058-25G TCIAV0058-5G
(S)-(+)-2-氨基-3-甲基-1-丁醇 ≥97.0% (by GC, titration analysis)
(S)-(+)-2-氨基-3-甲基-1-丁醇
分子式: C₅H₁₃NO
沸点: 88°C
溶点: 31…32 °C
密度: 0,92 g/cm³ (20 °C)
MDL编号: MFCD00064296
CAS号: 2026-48-4
EINECS: 217-975-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91decc88643caba7621b01aad31619dc96b82198&vid=91decc88643caba7621b01aad31619dc96b82198&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Specific rotation [a]20/D +15.0 to +19.0 deg (C=10, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR