EDTA tetrasodium salt dihydrate ≥98.0% (by titrimetric analysis)
供应商: TCI
|
Warning
|
Synonyms:
Tetrasodium ethylenediaminetetraacetate dihydrate, Edetic acid tetrasodium salt dihydrate, Kestranal® 4S 2 H2O, Ethylenedinitrilotetra-acetic acid tetrasodium salt dihydrate, Ethylenediaminetetra-acetic acid tetrasodium salt dihydrate, Ethylenediaminetetraacetic acid sodium hydrate, Tetrasodium Ethylenediaminetetraacetate Dihydrate, Kestranal® 4S dihydrate
E0099-25G
E0099-500G
TCIAE0099-25GEA
0
CNY
TCIAE0099-25G
TCIAE0099-500G
EDTA tetrasodium salt dihydrate ≥98.0% (by titrimetric analysis)
EDTA tetrasodium salt dihydrate
分子式:
C₁₀H₁₂N₂Na₄O₈·2H₂O 分子量: 416.2 g/mol 溶点: 300°C 存储条件: Ambient |
MDL编号:
MFCD00012460 CAS号: 10378-23-1 EINECS: 200-573-9 Merck Index: 13,03544 |
|
产品技术规格
Appearance | White powder to crystal |
Purity (Chelometric Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
pH | 10.0 to 12.0 (5 g/L, 25 deg-C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...