1-辛炔-3-醇 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
Oct-1-yn-3-ol
O0196-25ML
O0196-250ML
TCIAO0196-25MLEA
0
CNY
TCIAO0196-25ML
TCIAO0196-250ML
1-辛炔-3-醇 ≥98.0% (by GC)
1-辛炔-3-醇
分子式:
C₈H₁₄O 沸点: 83°C 溶点: 0 °C 密度: 0,864 g/cm³ (20 °C) 闪点: 63°C |
MDL编号:
MFCD00004588 CAS号: 818-72-4 EINECS: 212-455-4 ADR: 6.1,III |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...