(±)-3-Undecanol ≥94.0% (by GC)
供应商: TCI
Warning
Synonyms: (±)-3-Hydroxy undecane, 3-Undecanol
U0028-25ML U0028-5ML
TCIAU0028-25MLEA 0 CNY
TCIAU0028-25ML TCIAU0028-5ML
(±)-3-Undecanol ≥94.0% (by GC)
(±)-3-Undecanol
分子式: C₁₁H₂₄O
分子量: 172.31 g/mol
沸点: 132°C
闪点: 38°C
MDL编号: MFCD00046730
CAS号: 6929-08-4
EINECS: 230-054-2
UN号: 1987
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1859437cbbb8659828420bded8bcb632500294f&vid=e1859437cbbb8659828420bded8bcb632500294f&prid=web&clid=SAPDAM
现在订购
产品技术规格
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 94.0 %