DL-丙氨醇 ≥98.0% (通过GC,滴定分析)

供应商: TCI
Danger

A0942-25G A0942-5G
TCIAA0942-25GEA 0 CNY
TCIAA0942-25G TCIAA0942-5G
DL-丙氨醇 ≥98.0% (通过GC,滴定分析)
DL-丙氨醇
分子式: C₃H₉NO
沸点: 175°C
溶点: 8 °C
密度: 0,928 g/cm³ (20 °C)
闪点: 83°C
存储条件: Ambient
MDL编号: MFCD00008085
CAS号: 6168-72-5
EINECS: 228-207-3
UN号: 2735
ADR: 8,II
Merck Index: 14,00468
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81fc014e12b40b8199bfeff6364f386246918d73&vid=81fc014e12b40b8199bfeff6364f386246918d73&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Specific gravity (20/20) 0.9620 to 0.9660
Refractive index n20/D 1.4480 to 1.4520


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR