DL-Malic acid ≥99.0% (by titrimetric analysis)
供应商: TCI
|
Warning
|
Synonyms:
DL-Hydroxysuccinic acid, (±)-Malic acid, (±)-Hydroxy-1,4-butanedioic acid, (±)-Hydroxysuccinic acid, Hydroxybutanedioic acid, 2-Hydroxysuccinic acid
M0020-25G
M0020-500G
TCIAM0020-25GEA
0
CNY
TCIAM0020-25G
TCIAM0020-500G
DL-Malic acid ≥99.0% (by titrimetric analysis)
DL-Malic acid
分子式:
HOOCCH(OH)CH₂COOH 分子量: 134.09 g/mol 沸点: 306 存储条件: Ambient |
MDL编号:
MFCD00064212 CAS号: 617-48-1 EINECS: 230-022-8 Merck Index: 14,05707 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 129.0 to 134.0 deg-C |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...