2,3-二溴-1-丙醇 ≥95.0% (by GC)

供应商: TCI
Danger

Synonyms: 2,3-Dibromo-1-propanol, Glycerol alpha,beta-Dibromohydrin, alpha,beta-Dibromohydrin

D0204-25G D0204-500G
TCIAD0204-25GEA 0 CNY
TCIAD0204-25G TCIAD0204-500G
2,3-二溴-1-丙醇 ≥95.0% (by GC)
2,3-二溴-1-丙醇
分子式: HOCH₂BrCHCH₂Br
沸点: 99°C
溶点: 8 °C
密度: 2,1682 g/cm³ (0 °C)
MDL编号: MFCD00004699
CAS号: 96-13-9
EINECS: 202-480-9
UN号: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fe83d0694b943e8251e00aef964b2f3e3d491ed&vid=0fe83d0694b943e8251e00aef964b2f3e3d491ed&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR