2-溴丁酸 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Danger
|
Synonyms:
2-Bromobutanoic acid, (±)-2-Bromobutanoic acid
B0564-25G
B0564-500G
TCIAB0564-25GEA
0
CNY
TCIAB0564-25G
TCIAB0564-500G
2-溴丁酸 ≥98.0% (by GC, titration analysis)
2-溴丁酸
分子式:
H₃CCH₂BrCHCOOH 分子量: 167 g/mol 沸点: 214 溶点: –4°C |
MDL编号:
MFCD00004216 CAS号: 80-58-0 EINECS: 201-294-5 UN号: 3265 Merck Index: 14,01413 |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...