顺铂
供应商: TCI
|
Danger
|
Synonyms:
顺-二氯二氨基铂(II), 顺式-二胺二氯铂, 顺氯氨铂
D3371-1G
D3371-100MG
TCIAD3371-1GEA
0
CNY
TCIAD3371-1G
TCIAD3371-100MG
顺铂
顺铂
分子式:
H₆Cl₂N₂Pt 溶点: 270°C 密度: 3,738 g/cm³ (20 °C) |
MDL编号:
MFCD00011623 CAS号: 15663-27-1 EINECS: 239-733-8 UN号: 3288 ADR: 6.1,II Merck Index: 14,02317 |
|
产品技术规格
Appearance | Light orange to Yellow to Green powder to crystal |
Elemental analysis (Nitrogen) | 8.90 to 9.80 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...