三苯基乙酸 ≥97.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 三苯乙酸

T1585-25G T1585-5G
TCIAT1585-25GEA 0 CNY
TCIAT1585-25G TCIAT1585-5G
三苯基乙酸 ≥97.0% (by titrimetric analysis)
三苯基乙酸
分子式: C₂₀H₁₆O₂
沸点: 437 °C (1013 hPa)
溶点: 267…273 °C
密度: 1,27 g/cm³ (20 °C)
MDL编号: MFCD00004185
CAS号: 595-91-5
EINECS: 209-873-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3d01b35dff56b84e952364309c7dde7c19d9e5f&vid=a3d01b35dff56b84e952364309c7dde7c19d9e5f&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 97.0 %
Solubility in THF almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR