山梨酸乙酯 ≥98.0% (by GC)
供应商: TCI
Synonyms:
反,反-2,4-己二烯酸乙酯
S0055-25ML
S0055-500ML
TCIAS0055-25MLEA
0
CNY
TCIAS0055-25ML
TCIAS0055-500ML
山梨酸乙酯 ≥98.0% (by GC)
山梨酸乙酯
分子式:
C₈H₁₂O₂ 分子量: 140.18 g/mol 沸点: 76°C 闪点: 86°C |
MDL编号:
MFCD00009296 CAS号: 2396-84-1 EINECS: 219-258-2 |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...