四辛基溴化铵 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: Tetra-n-octylammonium bromide

T1603-10G T1603-25G
TCIAT1603-10GEA 0 CNY
TCIAT1603-10G TCIAT1603-25G
四辛基溴化铵 ≥98.0% (by titrimetric analysis)
四辛基溴化铵
分子式: C₃₂H₆₈BrN
分子量: 546.8 g/mol
溶点: 96
闪点: 100
MDL编号: MFCD00011863
CAS号: 14866-33-2
EINECS: 238-936-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e75d3e28c9f8d49b207d9626e571dc538baea35&vid=9e75d3e28c9f8d49b207d9626e571dc538baea35&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 96.0 to 99.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR