肉豆蔻酸甲酯 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: Myristic acid methyl ester, 十四酸甲酯

M0482-25ML M0482-500ML
TCIAM0482-25MLEA 0 CNY
TCIAM0482-25ML TCIAM0482-500ML
肉豆蔻酸甲酯 ≥98.0% (by GC)
肉豆蔻酸甲酯
分子式: C₁₅H₃₀O₂
分子量: 242.4 g/mol
沸点: 173°C
溶点: 18
闪点: 157°C
存储条件: Refrigerator
MDL编号: MFCD00008983
CAS号: 124-10-7
EINECS: 204-680-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27ca0087c2121f54002e6007a5ecf1697236682d&vid=27ca0087c2121f54002e6007a5ecf1697236682d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR