Tetradecanal ≥96.0% (by GC)

供应商: TCI
Warning

Synonyms: Tetradecanaldehyde, Tetradecanal, Myristaldehyde, Tetradecyl aldehyde, Myristyl aldehyde

T2696-1G T2696-5G
TCIAT2696-1GEA 0 CNY
TCIAT2696-1G TCIAT2696-5G
Tetradecanal ≥96.0% (by GC)
Tetradecanal
分子式: C₁₄H₂₈O
沸点: 166°C
溶点: 23°C
密度: 0,83 g/cm³ (20 °C)
存储条件: Freezer
MDL编号: MFCD00007019
CAS号: 124-25-4
EINECS: 204-692-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea970b8a8fc662629e1fe65267238a9ff46d0499&vid=ea970b8a8fc662629e1fe65267238a9ff46d0499&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR