丙酸丁酯 ≥99.0% (通过GC)
供应商: TCI
|
Warning
|
P0502-25ML
P0502-500ML
TCIAP0502-25MLEA
0
CNY
TCIAP0502-25ML
TCIAP0502-500ML
丙酸丁酯 ≥99.0% (通过GC)
丙酸丁酯
分子式:
C₇H₁₄O₂ 沸点: 146°C 溶点: –90 °C 密度: 0,88 g/cm³ (20 °C) 闪点: 42°C 存储条件: Ambient |
MDL编号:
MFCD00009448 CAS号: 590-01-2 EINECS: 209-669-5 UN号: 1914 ADR: 3,III Merck Index: 14,01587 |
|
产品技术规格
Appearance | Colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...