Polyethylene glycol 4,000
供应商: TCI
P0885-25G
P0885-500G
TCIAP0885-25GEA
0
CNY
TCIAP0885-25G
TCIAP0885-500G
Polyethylene glycol 4,000
Polyethylene glycol 4,000
分子式:
H(OCH₂CH₂)nOH 沸点: 溶点: 53…58 °C 密度: 1,13 g/cm³ (20 °C) 闪点: 250 °C 存储条件: Ambient |
MDL编号:
MFCD00081839 CAS号: 25322-68-3 EINECS: 500-038-2 Merck Index: 14,07568 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Average molecular weight | 2,600 to 3,800 |
Melting point | 56.0 to 60.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...