2,3,4,5,6-五氟苯甲酰胺 ≥99.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
2,3,4,5,6-Pentafluorobenzamide
P0929-25G
P0929-5G
TCIAP0929-25GEA
0
CNY
TCIAP0929-25G
TCIAP0929-5G
2,3,4,5,6-五氟苯甲酰胺 ≥99.0% (by GC)
2,3,4,5,6-五氟苯甲酰胺
分子式:
C₇H₂F₅NO 沸点: 272 °C (1013 hPa) 溶点: 151 °C |
MDL编号:
MFCD00007971 CAS号: 652-31-3 EINECS: 211-488-1 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 146.0 to 150.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...