硬脂酸甲酯 ≥97.0% (by GC)
供应商: TCI
Synonyms:
Stearic acid methyl ester, 十八酸甲酯
S0080-25G
S0080-500G
TCIAS0080-25GEA
0
CNY
TCIAS0080-25G
TCIAS0080-500G
硬脂酸甲酯 ≥97.0% (by GC)
硬脂酸甲酯
分子式:
CH₃(CH₂)₁₆CO₂CH₃ 分子量: 298.51 g/mol 沸点: 355 存储条件: Ambient |
MDL编号:
MFCD00009005 CAS号: 112-61-8 EINECS: 203-990-4 Merck Index: 12,08959 |
|
产品技术规格
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 97.0 % |
Melting point | 38.0 to 42.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...