硬脂酸酐 ≥97.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 十八酸酐

S0083-25G S0083-500G
TCIAS0083-25GEA 0 CNY
TCIAS0083-25G TCIAS0083-500G
硬脂酸酐 ≥97.0% (by titrimetric analysis)
硬脂酸酐
分子式: C₃₆H₇₀O₃
溶点: 72 °C
密度: 0,855 g/cm³ (60 °C)
存储条件: Freezer
MDL编号: MFCD00009004
CAS号: 638-08-4
EINECS: 211-318-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31df99206f84dd2a5991b1d62fcd260c22d3fc38&vid=31df99206f84dd2a5991b1d62fcd260c22d3fc38&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Morpholine Method) min. 97.0 %
Melting point 70.0 to 73.0 deg-C
Solubility in hot Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR