Nonacosanoic acid ≥95.0% (通过GC)
供应商: TCI
N0662-1G
N0662-5G
TCIAN0662-1GEA
0
CNY
TCIAN0662-1G
TCIAN0662-5G
Nonacosanoic acid ≥95.0% (通过GC)
Nonacosanoic acid
分子式:
C₂₉H₅₈O₂ |
MDL编号:
MFCD00057830 CAS号: 4250-38-8 EINECS: 224-210-9 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 88.0 to 92.0 deg-C |
Solubility in hot Toluene | within almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...