Hexyltrimethoxysilane ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: Trimethoxy(hexyl)silane

H0879-25ML H0879-500ML
TCIAH0879-25MLEA 0 CNY
TCIAH0879-25ML TCIAH0879-500ML
Hexyltrimethoxysilane ≥98.0% (by GC)
Hexyltrimethoxysilane
分子式: C₉H₂₂O₃Si
沸点: 203°C
闪点: 77°C
MDL编号: MFCD00039801
CAS号: 3069-19-0
EINECS: 221-331-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85f10f90167f2ceb4f5dea1fe272d5afe6617735&vid=85f10f90167f2ceb4f5dea1fe272d5afe6617735&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR