Hexyltriethoxysilane ≥97.0% (by GC)

供应商: TCI
Warning

Synonyms: Triethoxy(hexyl)silane

H1158-25ML H1158-500ML
TCIAH1158-25MLEA 0 CNY
TCIAH1158-25ML TCIAH1158-500ML
Hexyltriethoxysilane ≥97.0% (by GC)
Hexyltriethoxysilane
分子式: C₁₂H₂₈O₃Si
闪点: 95°C
MDL编号: MFCD00087744
CAS号: 18166-37-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c36a223a3ae9172d77b549f78f57c478af75936e&vid=c36a223a3ae9172d77b549f78f57c478af75936e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR