草酸二甲酯 ≥99.0% (by GC)

供应商: TCI
Warning

Synonyms: Oxalic acid dimethyl ester

O0080-25G O0080-500G
TCIAO0080-25GEA 0 CNY
TCIAO0080-25G TCIAO0080-500G
草酸二甲酯 ≥99.0% (by GC)
草酸二甲酯
分子式: C₄H₆O₄
分子量: 118.09 g/mol
沸点: 164°C
溶点: 54
闪点: 62
存储条件: Ambient
MDL编号: MFCD00008442
CAS号: 553-90-2
EINECS: 209-053-6
Merck Index: 14,06106
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd9626343cf0139c93b3e184c1e18509d2c1d717&vid=bd9626343cf0139c93b3e184c1e18509d2c1d717&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 53.0 to 57.0 deg-C
Solubility in EtOH very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR