草酸二丁酯 ≥99.0% (by GC)

供应商: TCI

Synonyms: Oxalic acid dibutyl ester

O0077-25ML O0077-500ML
TCIAO0077-25MLEA 0 CNY
TCIAO0077-25ML TCIAO0077-500ML
草酸二丁酯 ≥99.0% (by GC)
草酸二丁酯
分子式: C₁₀H₁₈O₄
分子量: 202.25 g/mol
沸点: 246°C
溶点: –29°C
闪点: 122°C
存储条件: Ambient
MDL编号: MFCD00009442
CAS号: 2050-60-4
EINECS: 218-092-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5df34b33dfb5687fcfb37123164dbcd717f6bdb&vid=f5df34b33dfb5687fcfb37123164dbcd717f6bdb&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR