二丙基二硫 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 二丙基二硫醚

D0932-25ML D0932-500ML
TCIAD0932-25MLEA 0 CNY
TCIAD0932-25ML TCIAD0932-500ML
二丙基二硫 ≥98.0% (by GC)
二丙基二硫
分子式: C₆H₁₄S₂
分子量: 150.31 g/mol
沸点: 193°C
溶点: -85.6
闪点: 82°C
存储条件: Ambient
MDL编号: MFCD00009378
CAS号: 629-19-6
EINECS: 211-079-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9923b9ae884e5970c04a5b8d93bc2777752eaae0&vid=9923b9ae884e5970c04a5b8d93bc2777752eaae0&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR