二苯乙酸 ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: 2,2-Diphenylacetic acid

D0869-25G D0869-500G
TCIAD0869-25GEA 0 CNY
TCIAD0869-25G TCIAD0869-500G
二苯乙酸 ≥98.0% (by HPLC, titration analysis)
二苯乙酸
分子式: C₁₄H₁₂O₂
沸点: 285 °C (1013 hPa)
溶点: 142…143 °C
密度: 1,258 g/cm³ (15 °C)
存储条件: Ambient
MDL编号: MFCD00004251
CAS号: 117-34-0
EINECS: 204-185-0
Merck Index: 14,03316
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fd00d90aa169575d080d96a1423236c561d7b7b&vid=0fd00d90aa169575d080d96a1423236c561d7b7b&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 146.0 to 149.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR