溴三甲基硅烷 ≥95.0% (by GC)
供应商: TCI
|
Danger
|
Synonyms:
三甲基溴化硅, 三甲基溴硅烷, TMBS
B1087-25ML
B1087-250ML
B1087-5ML
TCIAB1087-25MLEA
0
CNY
TCIAB1087-25ML
TCIAB1087-250ML
TCIAB1087-5ML
溴三甲基硅烷 ≥95.0% (by GC)
溴三甲基硅烷
分子式:
C₃H₉BrSi 分子量: 153.09 g/mol 沸点: 79°C 溶点: -43 闪点: 32°C 存储条件: Refrigerator |
MDL编号:
MFCD00000048 CAS号: 2857-97-8 EINECS: 220-672-0 UN号: 2920 ADR: 8 II |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...