7-甲氧基吲哚 ≥98.0% (通过GC)

供应商: TCI
Warning

M1751-1G M1751-5G
TCIAM1751-1GEA 0 CNY
TCIAM1751-1G TCIAM1751-5G
7-甲氧基吲哚 ≥98.0% (通过GC)
7-甲氧基吲哚
分子式: C₉H₉NO
沸点: 166°C
密度: 1,126 g/cm³ (25 °C)
存储条件: Freezer
MDL编号: MFCD00047203
CAS号: 3189-22-8
EINECS: 221-690-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8ecdf02b452a05069a9234ac6016f3ccc53d1b6&vid=b8ecdf02b452a05069a9234ac6016f3ccc53d1b6&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR