5-己烯-1-醇 ≥97.0% (by GC)
供应商: TCI
Warning
Synonyms: 5-己基-1-醇
H0653-10ML H0653-25ML H0653-250ML
TCIAH0653-10MLEA 0 CNY
TCIAH0653-10ML TCIAH0653-25ML TCIAH0653-250ML
5-己烯-1-醇 ≥97.0% (by GC)
5-己烯-1-醇
分子式: C₆H₁₂O
沸点: 157°C
溶点: –20 °C
密度: 0,84 g/cm³ (20 °C)
闪点: 47°C
存储条件: Ambient
MDL编号: MFCD00002981
CAS号: 821-41-0
EINECS: 212-477-4
UN号: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9de355d30db7aa1f4d55e2b0090d652420f2fb21&vid=9de355d30db7aa1f4d55e2b0090d652420f2fb21&prid=web&clid=SAPDAM
现在订购
产品技术规格
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %