6-氨基己酸 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: ε-氨基己酸, 6-氨己酸, EACA

A0312-25G A0312-500G
TCIAA0312-25GEA 0 CNY
TCIAA0312-25G TCIAA0312-500G
6-氨基己酸 ≥98.0% (by titrimetric analysis)
6-氨基己酸
分子式: C₆H₁₃NO₂
分子量: 131.17 g/mol
沸点: 257
溶点: 204°C
闪点: 207
存储条件: Ambient
MDL编号: MFCD00008238
CAS号: 60-32-2
EINECS: 200-469-3
Merck Index: 14,00432
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f909d2095137717168b2ae4926fdc4d1b8fdd55&vid=2f909d2095137717168b2ae4926fdc4d1b8fdd55&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR