吲哚-5-羧酸 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 1H-Indole-5-carboxylic acid

I0029-1G I0029-5G
TCIAI0029-1GEA 0 CNY
TCIAI0029-1G TCIAI0029-5G
吲哚-5-羧酸 ≥98.0% (by GC, titration analysis)
吲哚-5-羧酸
分子式: C₉H₇NO₂
分子量: 161.16 g/mol
存储条件: Freezer
MDL编号: MFCD00005678
CAS号: 1670-81-1
EINECS: 216-799-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ecd479462ebead7502e0a9b4731b51403c7d552&vid=2ecd479462ebead7502e0a9b4731b51403c7d552&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light red to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 209.0 to 214.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR