4-戊烯-1-醇 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
2-烯丙基乙醇
P1243-25ML
P1243-250ML
TCIAP1243-25MLEA
0
CNY
TCIAP1243-25ML
TCIAP1243-250ML
4-戊烯-1-醇 ≥98.0% (by GC)
4-戊烯-1-醇
分子式:
C₅H₁₀O 分子量: 86.13 g/mol 沸点: 138°C 闪点: 48°C 存储条件: Ambient |
MDL编号:
MFCD00002975 CAS号: 821-09-0 EINECS: 212-473-2 UN号: 1987 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...