5-氟吲哚 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 5-Fluoro-1H-indole

F0369-1G F0369-5G
TCIAF0369-1GEA 0 CNY
TCIAF0369-1G TCIAF0369-5G
5-氟吲哚 ≥98.0% (by GC)
5-氟吲哚
分子式: C₈H₆FN
分子量: 135.14 g/mol
沸点: 120°C
溶点: 46
存储条件: Refrigerator
MDL编号: MFCD00005671
CAS号: 399-52-0
EINECS: 206-917-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f55af34532e09d88fd6d7fd1b300de2e05f24b9e&vid=f55af34532e09d88fd6d7fd1b300de2e05f24b9e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 44.0 to 49.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR