5-氯-1-戊炔 ≥96.0% (by GC)
供应商: TCI
Danger
C1522-25ML C1522-5ML
TCIAC1522-25MLEA 0 CNY
TCIAC1522-25ML TCIAC1522-5ML
5-氯-1-戊炔 ≥96.0% (by GC)
5-氯-1-戊炔
分子式: C₅H₇Cl
分子量: 102.56 g/mol
沸点: 113°C
闪点: 15°C
存储条件: Freezer
MDL编号: MFCD00001014
CAS号: 14267-92-6
EINECS: 238-159-5
UN号: 1993
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc04264b4e710d457b33d5e008064043677215a0&vid=dc04264b4e710d457b33d5e008064043677215a0&prid=web&clid=SAPDAM
现在订购
产品技术规格
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %