异烟酸 ≥99.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: 吡啶-4-羧酸, 4-吡啶甲酸

I0207-25G I0207-500G
TCIAI0207-25GEA 0 CNY
TCIAI0207-25G TCIAI0207-500G
异烟酸 ≥99.0% (by HPLC, titration analysis)
异烟酸
分子式: C₆H₅NO₂
分子量: 123.11 g/mol
沸点: 396
溶点: 315°C
存储条件: Ambient
MDL编号: MFCD00006429
CAS号: 55-22-1
EINECS: 200-228-2
Merck Index: 14,05187
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d3a1153ec3f01fec0c4c009a47f21ccd7f26c28&vid=9d3a1153ec3f01fec0c4c009a47f21ccd7f26c28&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Neutralization titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR