4-甲基-2-戊醇 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
甲基异丁基甲醇
M0386-25ML
M0386-500ML
TCIAM0386-25MLEA
0
CNY
TCIAM0386-25ML
TCIAM0386-500ML
4-甲基-2-戊醇 ≥98.0% (by GC)
4-甲基-2-戊醇
分子式:
C₆H₁₄O 分子量: 102.18 g/mol 沸点: 132°C 溶点: –90°C 闪点: 43°C 存储条件: Ambient |
MDL编号:
MFCD00004550 CAS号: 108-11-2 EINECS: 203-551-7 UN号: 2053 ADR: 3 III |
|
产品技术规格
Purity (GC) | min. 98.0 % |
Appearance | Colorless to Almost colorless clear liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...