4-溴-2-氯吡啶 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
B2951-1G
B2951-5G
TCIAB2951-1GEA
0
CNY
TCIAB2951-1G
TCIAB2951-5G
4-溴-2-氯吡啶 ≥98.0% (by GC)
4-溴-2-氯吡啶
分子式:
C₅H₃BrClN 分子量: 192.44 g/mol 沸点: 70°C |
MDL编号:
MFCD03840756 CAS号: 73583-37-6 |
|
产品技术规格
Appearance | White or Colorless to Yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Freezing point | 18.0 to 22.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...