4-氨基-2-氯吡啶
供应商: TCI
|
Warning
|
Synonyms:
2-Chloro-4-aminopyridine
A0895-1G
A0895-25G
A0895-5G
TCIAA0895-1GEA
0
CNY
TCIAA0895-1G
TCIAA0895-25G
TCIAA0895-5G
4-氨基-2-氯吡啶
4-氨基-2-氯吡啶
分子式:
C₅H₅ClN₂ 分子量: 128.56 g/mol 沸点: 300 |
MDL编号:
MFCD00060089 CAS号: 14432-12-3 EINECS: 238-403-0 |
|
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 90.0 to 94.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...